2-klorbensofenon ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: o-Chlorobenzophenone, (2-Chlorophenyl)(phenyl)methanone

C1150-25G C1150-500G C1150-100G
TCIAC1150-25GEA 270 NOK
TCIAC1150-25G TCIAC1150-500G TCIAC1150-100G
2-klorbensofenon ≥98.0% (by GC)
2-klorbensofenon
Formel: C₁₃H₉ClO
MW: 216,67 g/mol
Kokepunkt: 180°C
Smeltepunkt: 76 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00000558
CAS-nummer: 5162-03-8
EINECS: 225-936-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2789bc0f3395649c652adf80d4bc1e1e45fca8a8&vid=2789bc0f3395649c652adf80d4bc1e1e45fca8a8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR