2-Amino-6-klorpurin ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI

Synonymer: 6-Chloroguanine, 6-Chloro-7H-purin-2-ylamine

A1407-1G A1407-5G
TCIAA1407-1GEA 220 NOK
TCIAA1407-1G TCIAA1407-5G
2-Amino-6-klorpurin ≥98.0% (by HPLC, titration analysis)
2-Amino-6-chloropurine
Formel: C₅H₄ClN₅
MW: 169,57 g/mol
Smeltepunkt: 300 °C
MDL nummer: MFCD00075252
CAS-nummer: 10310-21-1
EINECS: 233-686-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4bf2b9e0f616d98a54d88681c53703f87b35bb4&vid=f4bf2b9e0f616d98a54d88681c53703f87b35bb4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR