2-Amino-3,5-dichloropyridine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 3,5-Dichloro-2-pyridylamine, 3,5-Dichloropyridin-2-amine

A1502-25G A1502-5G
TCIAA1502-25GEA 1100 NOK
TCIAA1502-25G TCIAA1502-5G
2-Amino-3,5-dichloropyridine ≥98.0% (by GC, titration analysis)
2-Amino-3,5-dichloropyridine
Formel: C₅H₄Cl₂N₂
MW: 163,01 g/mol
Smeltepunkt: 79…83 °C
MDL nummer: MFCD00006313
CAS-nummer: 4214-74-8
EINECS: 224-143-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f97d5964ffd01eb139f57126ea001a0b8df0ba75&vid=f97d5964ffd01eb139f57126ea001a0b8df0ba75&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR