3,4-Dicyanoanilin ≥98.0% (by total nitrogen basis)
Leverandør: TCI EUROPE
|
Danger
|
TCIAA1166-5GEA
1550
NOK
TCIAA1166-5G
TCIAA1166-25G
3,4-Dicyanoanilin ≥98.0% (by total nitrogen basis)
3,4-Dicyanoanilin
Formel:
C₈H₅N₃ MW: 143,15 g/mol Smeltepunkt: 180…182 °C |
MDL nummer:
MFCD00041741 CAS-nummer: 56765-79-8 EINECS: 260-370-6 FN: 2811 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 179.0 to 182.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...