Dibenzosuberone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-Dibenzo[b,f]cycloheptanone, 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one, 5-Dibenzosuberone, Dibenzo[b,f]cycloheptan-1-one

D1801-25G D1801-100G
TCIAD1801-25GEA 230 NOK
TCIAD1801-25G TCIAD1801-100G
Dibenzosuberone ≥98.0% (by GC)
Dibenzosuberone
Formel: C₁₅H₁₂O
MW: 208,26 g/mol
Kokepunkt: 148°C
Smeltepunkt: 31…34 °C
Tetthet: 1,1635 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003587
CAS-nummer: 1210-35-1
EINECS: 214-912-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a17841e9bcc9933a4928d0fb9903407ca64d6155&vid=a17841e9bcc9933a4928d0fb9903407ca64d6155&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR