3',4'-Dimethoxyacetophenone ≥98.0% (by GC)

Leverandør: TCI

Synonymer: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone

D1878-25G D1878-500G D1878-100G
TCIAD1878-25GEA 405 NOK
TCIAD1878-25G TCIAD1878-500G TCIAD1878-100G
3',4'-Dimethoxyacetophenone ≥98.0% (by GC)
3',4'-Dimethoxyacetophenone
Formel: C₁₀H₁₂O₃
MW: 180,2 g/mol
Kokepunkt: 161°C
Smeltepunkt: 48…49 °C
MDL nummer: MFCD00008737
CAS-nummer: 1131-62-0
EINECS: 214-468-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f91fc112a2d1861343113f6d16d6d2d2ecc7f75&vid=8f91fc112a2d1861343113f6d16d6d2d2ecc7f75&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 52.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR