3',4'-Dihydroxyacetophenone ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 1-(3,4-Dihydroxyphenyl)ethan-1-one, Acetopyrocatechol

D2345-25G D2345-5G
TCIAD2345-25GEA 2975 NOK
TCIAD2345-25G TCIAD2345-5G
3',4'-Dihydroxyacetophenone ≥98.0% (by GC)
3',4'-Dihydroxyacetophenone
Formel: C₈H₈O₃
MW: 152,15 g/mol
MDL nummer: MFCD00016439
CAS-nummer: 1197-09-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7d9cf6a6280e72e7e4bcc2a06b7fc5f44937b700&vid=7d9cf6a6280e72e7e4bcc2a06b7fc5f44937b700&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 118.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR