p-Benzoquinon ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
p-Quinone, Quinone, 1,4-Cyclohexadiene-3,6-dione, 1,4-Benzoquinone
B0089-25G
B0089-500G
B0089-100G
TCIAB0089-25GEA
245
NOK
TCIAB0089-25G
TCIAB0089-500G
TCIAB0089-100G
p-Benzoquinon ≥98.0% (by titrimetric analysis)
p-Benzoquinon
Formel:
C₆H₄O₂ MW: 108,1 g/mol Kokepunkt: ~ 174 °C (1013 hPa) Smeltepunkt: 112…114 °C Tetthet: 1,318 g/cm³ (20 °C) Flash punkt: 77 °C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00001591 CAS-nummer: 106-51-4 EINECS: 203-405-2 FN: 2587 ADR: 6.1,II Merck Indeks: 14,08074 |
Spesifikasjon Testresultater
Appearance | Light yellow to Yellow to Orange powder to crystaline |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 112.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...