Methyl-p-benzoquinone ≥99.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Methylcyclohexa-2,5-diene-1,4-dione, Methylquinone, Methyl-1,4-benzoquinone
M0159-25G
M0159-5G
TCIAM0159-25GEA
3475
NOK
TCIAM0159-25G
TCIAM0159-5G
Methyl-p-benzoquinone ≥99.0% (by GC, titration analysis)
Methyl-p-benzoquinone
Formel:
C₇H₆O₂ MW: 122,12 g/mol Kokepunkt: 187 °C (1013 hPa) Smeltepunkt: 65…67 °C Tetthet: 1,08 g/cm³ (75.5 °C) |
MDL nummer:
MFCD00001603 CAS-nummer: 553-97-9 EINECS: 209-056-2 |
Spesifikasjon Testresultater
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Iodometric Titration) | min. 99.0 % |
Melting point | 66.0 to 69.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...