Tetramethyl-p-benzoquinone ≥98.0% (by GC)
Leverandør: TCI
Warning
Synonymer: 2,3,5,6-Tetramethylcyclohexa-2,5-diene-1,4-dione, 2,3,5,6-Tetramethyl-1,4-benzoquinone, Tetramethylquinone, Tetramethyl-1,4-benzoquinone, Duroquinone
T0672-1G T0672-25G T0672-5G
TCIAT0672-1GEA 715 NOK
TCIAT0672-1G TCIAT0672-25G TCIAT0672-5G
Tetramethyl-p-benzoquinone ≥98.0% (by GC)
Tetramethyl-p-benzoquinone
Formel: C₁₀H₁₂O₂
MW: 164,2 g/mol
Kokepunkt: 282 °C (1013 hPa)
Smeltepunkt: 111…113 °C
MDL nummer: MFCD00001604
CAS-nummer: 527-17-3
EINECS: 208-409-8
Merck Indeks: 14,03470
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6d562bbf09820714a7cfc771ee64b67bc447eeb&vid=f6d562bbf09820714a7cfc771ee64b67bc447eeb&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 112.0 to 114.0 deg-C
Solubility in hot Methanol almost transparency