Butyramide ≥98.0% (by GC, total nitrogen)
Leverandør: TCI
B0809-25G
B0809-500G
TCIAB0809-25GEA
220
NOK
TCIAB0809-25G
TCIAB0809-500G
Butyramide ≥98.0% (by GC, total nitrogen)
Butyramide
Formel:
C₄H₉NO MW: 87,12 g/mol Kokepunkt: 216°C Smeltepunkt: 115…116 °C Tetthet: 1,027 g/cm³ (20 °C) |
MDL nummer:
MFCD00041894 CAS-nummer: 541-35-5 EINECS: 208-776-4 Merck Indeks: 14,01592 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 114.0 to 117.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...