Bis(2-etylheksyl)-ftalat ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
Di-iso-octyl phthalate, Dioctyl phthalate, Di-(2-ethylhexyl) phthalate, DEHP, Di(2-ethylhexyl) phthalate, Phthalic acid bis(2-ethylhexyl) ester
P0297-25G
P0297-500G
TCIAP0297-25GEA
230
NOK
TCIAP0297-25G
TCIAP0297-500G
Bis(2-etylheksyl)-ftalat ≥98.0% (by GC)
Bis(2-etylheksyl)-ftalat
Formel:
C₂₄H₃₈O₄ MW: 390.56 g/mol Kokepunkt: 361°C Smeltepunkt: -50 Flash punkt: 218°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00009493 CAS-nummer: 117-81-7 EINECS: 204-211-0 FN: 3082 ADR: 9 III Merck Indeks: 14,02864 |
|
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |