2,2-Dimethylbutyric acid ≥96.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
2,2-Dimethylbutanoic acid
D2429-25ML
D2429-500ML
TCIAD2429-25MLEA
230
NOK
TCIAD2429-25ML
TCIAD2429-500ML
2,2-Dimethylbutyric acid ≥96.0% (by GC, titration analysis)
2,2-Dimethylbutyric acid
Formel:
C₆H₁₂O₂ MW: 116,16 g/mol Kokepunkt: 186°C Smeltepunkt: –13 °C Tetthet: 0,9449 g/cm³ (0 °C) Flash punkt: 79°C |
MDL nummer:
MFCD00004200 CAS-nummer: 595-37-9 EINECS: 209-865-0 FN: 3265 ADR: 8,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...