Picolinic acid ≥99.0% (by titrimetric analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
Pyridine-2-carboxylic acid, 2-Picolinic acid, 2-Pyridinecarboxylic acid
P0421-25G
P0421-500G
P0421-100G
TCIAP0421-25GEA
260
NOK
TCIAP0421-25G
TCIAP0421-500G
TCIAP0421-100G
Picolinic acid ≥99.0% (by titrimetric analysis)
Picolinic acid
Formel:
C₆H₅NO₂ MW: 123,11 g/mol Kokepunkt: 293 °C (1013 hPa) Smeltepunkt: 134…137 °C Tetthet: 1,49 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00006293 CAS-nummer: 98-98-6 EINECS: 202-719-7 Merck Indeks: 14,07403 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 138.0 to 142.0 deg-C |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...