4-Pentenoic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: Allylacetic acid, Pent-4-enoic acid

P0645-25ML P0645-100ML
TCIAP0645-25MLEA 650 NOK
TCIAP0645-25ML TCIAP0645-100ML
4-Pentenoic acid ≥98.0% (by GC, titration analysis)
4-Pentenoic acid
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokepunkt: 182°C
Smeltepunkt: –18 °C
Tetthet: 0,98 g/cm³ (20 °C)
Flash punkt: 93°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004408
CAS-nummer: 591-80-0
EINECS: 209-732-7
FN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cac82daf62f92cc6ac982ba5c959e9a499a87c0a&vid=cac82daf62f92cc6ac982ba5c959e9a499a87c0a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR