3-(3,4-Dimethoxyphenyl)propionic acid ≥98.0% (by titrimetric analysis)

Leverandør: TCI

Synonymer: 3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid

D1953-10G D1953-25G
TCIAD1953-10GEA 570 NOK
TCIAD1953-10G TCIAD1953-25G
3-(3,4-Dimethoxyphenyl)propionic acid ≥98.0% (by titrimetric analysis)
3-(3,4-Dimethoxyphenyl)propionic acid
Formel: C₁₁H₁₄O₄
MW: 210.23 g/mol
Kokepunkt: 344
Smeltepunkt: 102
MDL nummer: MFCD00002774
CAS-nummer: 2107-70-2
EINECS: 218-288-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8afd1d24d6090c32dedcc2980ce90396d1ef3312&vid=8afd1d24d6090c32dedcc2980ce90396d1ef3312&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 98.0 to 101.0 deg-C
Solubility in Methanol very faint turbidity

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR