(2-Hydroxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Hydroxyphenylacetic acid
H0340-25G
H0340-5G
TCIAH0340-25GEA
635
NOK
TCIAH0340-25G
TCIAH0340-5G
(2-Hydroxyphenyl)acetic acid ≥98.0% (by GC, titration analysis)
(2-Hydroxyphenyl)acetic acid
Formel:
(C₆H₄OH)CH₂COOH MW: 152,15 g/mol Kokepunkt: 328 °C (1013 hPa) Smeltepunkt: 145…146 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004323 CAS-nummer: 614-75-5 EINECS: 210-393-2 |
Spesifikasjon Testresultater
Appearance | White to Light yellow to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 143.0 to 148.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...