2-(Trifluoromethyl)cinnamic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
2-(Trifluoromethyl)cinnamicacid
T2608-25G
T2608-5G
TCIAT2608-25GEA
2075
NOK
TCIAT2608-25G
TCIAT2608-5G
2-(Trifluoromethyl)cinnamic acid ≥98.0% (by GC, titration analysis)
2-(Trifluoromethyl)cinnamic acid
Formel:
C₁₀H₇F₃O₂ MW: 216,16 g/mol Kokepunkt: 292 °C (1013 hPa) Smeltepunkt: 199 °C |
MDL nummer:
MFCD00004381 CAS-nummer: 2062-25-1 EINECS: 218-168-0 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 201.0 to 207.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...