2-(Trifluoromethyl)thiophenol ≥97.0% (by GC, titration analysis)
Leverandør: TCI
|
Danger
|
Synonymer:
2-(Trifluoromethyl)benzene-1-thiol, 2-(Trifluoromethyl)benzenethiol, 2-Mercaptobenzotrifluoride
T2519-1G
T2519-5G
TCIAT2519-1GEA
525
NOK
TCIAT2519-1G
TCIAT2519-5G
2-(Trifluoromethyl)thiophenol ≥97.0% (by GC, titration analysis)
2-(Trifluoromethyl)thiophenol
Formel:
C₇H₅F₃S MW: 178,18 g/mol Kokepunkt: 64°C Flash punkt: 68°C |
MDL nummer:
MFCD00068896 CAS-nummer: 13333-97-6 FN: 2810 ADR: 6.1,III |
Spesifikasjon Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...