6-Nitro-2,3-xylidine ≥98.0% (by GC)

Leverandør: TCI
Danger

N0500-25G N0500-500G
TCIAN0500-25GEA 435 NOK
TCIAN0500-25G TCIAN0500-500G
6-Nitro-2,3-xylidine ≥98.0% (by GC)
6-Nitro-2,3-xylidine
Formel: C₈H₁₀N₂O₂
Smeltepunkt: 120 °C
MDL nummer: MFCD00010148
CAS-nummer: 59146-96-2
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2f92da5462a87caf2096968c0192da1776fe9b8&vid=d2f92da5462a87caf2096968c0192da1776fe9b8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Orange to Brown to Dark red powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR