2-Nitrobenzyl alcohol ≥98.0% (by GC)
Leverandør: TCI
Warning
Synonymer: (2-Nitrophenyl)methanol
N0179-25G N0179-250G
TCIAN0179-25GEA 420 NOK
TCIAN0179-25G TCIAN0179-250G
2-Nitrobenzyl alcohol ≥98.0% (by GC)
2-Nitrobenzyl alcohol
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Kokepunkt: 168°C
Smeltepunkt: 71…72 °C
MDL nummer: MFCD00007186
CAS-nummer: 612-25-9
EINECS: 210-302-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e0f8ce89c8e8a062dcd33cfd5b0d7d42a083cc5&vid=9e0f8ce89c8e8a062dcd33cfd5b0d7d42a083cc5&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 74.0 deg-C
Solubility in Methanol very faint turbidity