Diethyl sulphite ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
D0528-25G
D0528-500G
TCIAD0528-25GEA
435
NOK
TCIAD0528-25G
TCIAD0528-500G
Diethyl sulphite ≥98.0% (by GC)
Diethyl sulphite
Formel:
C₄H₁₀O₃S MW: 138,19 g/mol Kokepunkt: 157°C Tetthet: 1,08 g/cm³ (20 °C) Flash punkt: 49°C |
MDL nummer:
MFCD00009098 CAS-nummer: 623-81-4 EINECS: 210-815-5 FN: 1993 ADR: 3,III |
Spesifikasjon Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...