Trifenylfosfat ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Triphenylphosphate, TPP
P0272-25G
P0272-500G
TCIAP0272-25GEA
220
NOK
TCIAP0272-25G
TCIAP0272-500G
Trifenylfosfat ≥99.0% (by GC)
Trifenylfosfat
Formel:
C₁₈H₁₅O₄P MW: 326,29 g/mol Kokepunkt: 220°C Smeltepunkt: 50…51 °C Tetthet: 1,33 g/cm³ (20 °C) Flash punkt: 223 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00003031 CAS-nummer: 115-86-6 EINECS: 204-112-2 FN: 3077 ADR: 9,III Merck Indeks: 14,09742 |
Spesifikasjon Testresultater
Appearance | White to Almost white lump |
Purity (GC) | min. 99.0 % |
Melting point | 49.0 to 52.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...