Maleinsyreanhydrid ≥99.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 2,5-Furanedione

M0005-25G M0005-500G
TCIAM0005-25GEA 230 NOK
TCIAM0005-25G TCIAM0005-500G
Maleinsyreanhydrid ≥99.0% (by GC, titration analysis)
Maleinsyreanhydrid
Formel: C₄H₂O₃
MW: 98,06 g/mol
Kokepunkt: 199°C
Smeltepunkt: 51…53 °C
Tetthet: 1,48 g/cm³ (20 °C)
Flash punkt: 103 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005518
CAS-nummer: 108-31-6
EINECS: 203-571-6
FN: 2215
ADR: 8,III
Merck Indeks: 14,05704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9abd3e41098c808e06c986c5d87d1ba8b2ac52fd&vid=9abd3e41098c808e06c986c5d87d1ba8b2ac52fd&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White pellet
Purity (GC) min. 99.0 %
Purity (Methanolysis Method) min. 99.0 %
Melting point 52.0 to 56.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR