1,1-Diphenylethanol ≥98.0% (by GC)

Leverandør: TCI

Synonymer: α-Methylbenzhydryl alcohol

D0295-25G D0295-5G
TCIAD0295-25GEA 1500 NOK
TCIAD0295-25G TCIAD0295-5G
1,1-Diphenylethanol ≥98.0% (by GC)
1,1-Diphenylethanol
Formel: (C₆H₅)₂C(OH)CH₃
MW: 198.26 g/mol
Kokepunkt: 155°C
MDL nummer: MFCD00004450
CAS-nummer: 599-67-7
EINECS: 209-970-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d18c777513acf3bdea37170d7128a1ba221822f1&vid=d18c777513acf3bdea37170d7128a1ba221822f1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 79.0 to 82.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR