4-tert-Butylpyrokatekol ≥98.0% (by GC)

Leverandør: TCI
Danger

Synonymer: 4-(tert-Butyl)benzene-1,2-diol, 4-tert-Butylcatechol

B0739-25G B0739-500G
TCIAB0739-25GEA 220 NOK
TCIAB0739-25G TCIAB0739-500G
4-tert-Butylpyrokatekol ≥98.0% (by GC)
4-tert-Butylpyrokatekol
Formel: C₁₀H₁₄O₂
MW: 166,22 g/mol
Kokepunkt: 285 °C (1013 hPa)
Smeltepunkt: 53 °C
Tetthet: 1,05 g/cm³ (60 °C)
Flash punkt: 129 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00002201
CAS-nummer: 98-29-3
EINECS: 202-653-9
FN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e17cca7b3366c7ec4a08328cc82bc55f5384626a&vid=e17cca7b3366c7ec4a08328cc82bc55f5384626a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light red powder to lump
Purity (GC) min. 98.0 %
Melting point 53.0 to 58.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR