2-Methyl-2-propanethiol ≥98.0% (by GC)
Leverandør: TCI
|
Danger
|
Synonymer:
2-Methyl-2-propanethiol, tert-Butyl Mercaptan, tert-Butyl mercaptan, tert-Butylthiol
M0405-25ML
M0405-500ML
TCIAM0405-25MLEA
260
NOK
TCIAM0405-25ML
TCIAM0405-500ML
2-Methyl-2-propanethiol ≥98.0% (by GC)
2-Methyl-2-propanethiol
Formel:
C₄H₁₀S MW: 90,19 g/mol Kokepunkt: 64°C Smeltepunkt: –1°C Tetthet: 0,78888 g/cm³ (20 °C) Flash punkt: –26°C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00004857 CAS-nummer: 75-66-1 EINECS: 200-890-2 FN: 2347 ADR: 3,II Merck Indeks: 14,01579 |
Spesifikasjon Testresultater
Appearance | Colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...