2-Pyridintiol ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Warning

Synonymer: Pyridine-2-thiol, 2-Mercaptopyridine

M0246-100G M0246-25G
TCIAM0246-100GEA 1850 NOK
TCIAM0246-100G TCIAM0246-25G
2-Pyridintiol ≥98.0% (by HPLC, titration analysis)
2-Pyridintiol
Formel: C₅H₅NS
MW: 111,17 g/mol
Kokepunkt: 259 °C (1013 hPa)
Smeltepunkt: 132,5…133.5 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00006285
CAS-nummer: 2637-34-5
EINECS: 220-131-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4418729de18cb74ae4f1f4454b13aafcb1237312&vid=e981567863e2fd1c42f7c63bf3bd153e1335d139&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Amber powder to crystaline
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 127.0 to 132.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR