o-Terfenyl ≥99.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
2-Phenylbiphenyl, 1,2-Terphenyl, 2-Phenyl-1,1'-biphenyl, 1,2-Diphenylbenzene, 1,1:2,1-Terphenyl
T0019-25G
T0019-500G
T0019-100G
TCIAT0019-25GEA
515
NOK
TCIAT0019-25G
TCIAT0019-500G
TCIAT0019-100G
o-Terfenyl ≥99.0% (by GC)
o-Terfenyl
Formel:
C₁₈H₁₄ MW: 230,31 g/mol Kokepunkt: 332°C Smeltepunkt: 54…56 °C Tetthet: 1,1 g/cm³ (20 °C) Flash punkt: 163 °C Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00003055 CAS-nummer: 84-15-1 EINECS: 201-517-6 FN: 3077 ADR: 9,III |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 56.0 to 59.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...