2-Methylbenzimidazole ≥98.0% (by GC, titration analysis)
Leverandør: TCI
Warning
Synonymer: 2-Methyl-1H-benzo[d]imidazole, 2-Methyl-1H-benzimidazole
M0286-25G M0286-250G
TCIAM0286-25GEA 300 NOK
TCIAM0286-25G TCIAM0286-250G
2-Methylbenzimidazole ≥98.0% (by GC, titration analysis)
2-Methylbenzimidazole
Formel: C₈H₈N₂
MW: 132,16 g/mol
Kokepunkt: 339 °C (1013 hPa)
Smeltepunkt: 175 °C
MDL nummer: MFCD00005598
CAS-nummer: 615-15-6
EINECS: 210-411-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc3985c69babef7be38ad51f6e636025dae6b995&vid=bc3985c69babef7be38ad51f6e636025dae6b995&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 174.0 to 178.0 deg-C