2-Methylquinoxaline ≥98.0% (by GC)

Leverandør: TCI
Warning

M0580-25G M0580-5G
TCIAM0580-25GEA 1300 NOK
TCIAM0580-25G TCIAM0580-5G
2-Methylquinoxaline ≥98.0% (by GC)
2-Methylquinoxaline
Formel: C₉H₈N₂
MW: 144,18 g/mol
Kokepunkt: 245°C
Tetthet: 1,11 g/cm³ (20 °C)
Flash punkt: 107°C
MDL nummer: MFCD00006727
CAS-nummer: 7251-61-8
EINECS: 230-664-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba4756b85835c7ce64b84438c4ff4a0c22f91b4b&vid=ba4756b85835c7ce64b84438c4ff4a0c22f91b4b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR