1-Methylindole ≥96.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
N-Methylindole, 1-Methyl-1H-indole
M0561-25G
M0561-5G
TCIAM0561-25GEA
905
NOK
TCIAM0561-25G
TCIAM0561-5G
1-Methylindole ≥96.0% (by GC)
1-Methylindole
Formel:
C₉H₉N MW: 131,18 g/mol Kokepunkt: 246°C Smeltepunkt: 94…95 °C Tetthet: 1,048 g/cm³ (20 °C) |
MDL nummer:
MFCD00005800 CAS-nummer: 603-76-9 EINECS: 210-057-5 ADR: 9, |
Spesifikasjon Testresultater
Appearance | Colorless to Yellow to Orange clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...