Triphenylene ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 9,10-Benzophenanthrene, Isochrysene

T0513-1G T0513-100MG
TCIAT0513-1GEA 1050 NOK
TCIAT0513-1G TCIAT0513-100MG
Triphenylene ≥96.0% (by GC)
Triphenylene
Formel: C₁₈H₁₂
MW: 228,29 g/mol
Kokepunkt: 425°C
Smeltepunkt: 199 °C
Tetthet: 1,302 g/cm³ (20 °C)
MDL nummer: MFCD00001108
CAS-nummer: 217-59-4
EINECS: 205-922-9
ADR: 9,III
Merck Indeks: 14,09740
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=70150a32e28ffeea4b05475857393c4e5a78b30b&vid=70150a32e28ffeea4b05475857393c4e5a78b30b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 96.0 %
Melting point 196.0 to 200.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR