5-Bromo-2-furaldehyde ≥98.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

B1954-25G B1954-5G
TCIAB1954-25GEA 1575 NOK
TCIAB1954-25G TCIAB1954-5G
5-Bromo-2-furaldehyde ≥98.0% (by GC)
5-Bromo-2-furaldehyde
Formel: C₅H₃BrO₂
MW: 174,98 g/mol
Kokepunkt: 112°C
Smeltepunkt: 74…76 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00159501
CAS-nummer: 1899-24-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbaf42871bfdbda09c76b08fae3157bc0c1ef9d8&vid=dbaf42871bfdbda09c76b08fae3157bc0c1ef9d8&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 80.0 to 84.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR