(3-Bromophenyl)acetic acid ≥95.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 2-(3-Bromophenyl)acetic acid

B1927-25G B1927-5G
TCIAB1927-25GEA 1875 NOK
TCIAB1927-25G TCIAB1927-5G
(3-Bromophenyl)acetic acid ≥95.0% (by GC, titration analysis)
(3-Bromophenyl)acetic acid
Formel: (C₆H₄Br)CH₂COOH
MW: 215,05 g/mol
Kokepunkt: 308 °C (1013 hPa)
Smeltepunkt: 102…103 °C
MDL nummer: MFCD00004330
CAS-nummer: 1878-67-7
EINECS: 217-522-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6da73bf687788749dc0eb621bfdcab6f09595e5a&vid=6da73bf687788749dc0eb621bfdcab6f09595e5a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %
Melting point 98.0 to 104.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR