2-Bromo-5-iodopyridine ≥97.0% (by GC)

Leverandør: TCI
Warning

B3765-25G B3765-5G
TCIAB3765-25GEA 2025 NOK
TCIAB3765-25G TCIAB3765-5G
2-Bromo-5-iodopyridine ≥97.0% (by GC)
2-Bromo-5-iodopyridine
Formel: C₅H₃BrIN
MW: 283,89 g/mol
MDL nummer: MFCD03095201
CAS-nummer: 73290-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=516c10e839934ead051cdaa1b1ab81eecb6d629b&vid=516c10e839934ead051cdaa1b1ab81eecb6d629b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 121.0 to 125.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR