4-Bromo-2,6-xylidine ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Warning

Synonymer: 4-Bromo-2,6-dimethylaniline

B1909-100G B1909-25G
TCIAB1909-100GEA 3925 NOK
TCIAB1909-100G TCIAB1909-25G
4-Bromo-2,6-xylidine ≥98.0% (by GC, titration analysis)
4-Bromo-2,6-xylidine
Formel: C₈H₁₀BrN
MW: 200,08 g/mol
Kokepunkt: 120 °C (0,2 -rr)
Smeltepunkt: 50 °C
MDL nummer: MFCD00007826
CAS-nummer: 24596-19-8
EINECS: 246-337-9
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f9c739281cbc21d1e71030ca161d4d10f3a6c61&vid=4f9c739281cbc21d1e71030ca161d4d10f3a6c61&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Brown to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 48.0 to 52.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR