2-Amino-5-bromo-3-nitropyridine ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: 5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine

A1556-25G A1556-5G
TCIAA1556-25GEA 1475 NOK
TCIAA1556-25G TCIAA1556-5G
2-Amino-5-bromo-3-nitropyridine ≥97.0% (by GC)
2-Amino-5-bromo-3-nitropyridine
Formel: C₅H₄BrN₃O₂
MW: 218,01 g/mol
Smeltepunkt: 215…216 °C
MDL nummer: MFCD00047441
CAS-nummer: 6945-68-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76332cc30e8182328521d614b6df9806d2b28bfc&vid=76332cc30e8182328521d614b6df9806d2b28bfc&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 97.0 %
Melting point 207.0 to 212.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR