6-klorpurin ≥98.0% (by titrimetric analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine
C0278-25G
C0278-5G
TCIAC0278-25GEA
2725
NOK
TCIAC0278-25G
TCIAC0278-5G
6-klorpurin ≥98.0% (by titrimetric analysis)
6-klorpurin
Formel:
C₅H₃ClN₄ MW: 154,56 g/mol Kokepunkt: 185 - 192 °C (2 -rr) Smeltepunkt: 178…179 °C Lagringstemperatur: Kjøleskap |
MDL nummer:
MFCD00075825 CAS-nummer: 87-42-3 EINECS: 201-745-6 Merck Indeks: 14,02161 |
Spesifikasjon Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Solubility in hot Water | within almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...