3-Chloropropionic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Danger

Synonymer: 3-Chloropropanoic acid

C0276-25G C0276-500G
TCIAC0276-25GEA 220 NOK
TCIAC0276-25G TCIAC0276-500G
3-Chloropropionic acid ≥98.0% (by GC, titration analysis)
3-Chloropropionic acid
Formel: C₃H₅ClO₂
MW: 108,52 g/mol
Kokepunkt: 204 °C (1013 hPa)
Smeltepunkt: 37…40 °C
Tetthet: 1,26 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00002764
CAS-nummer: 107-94-8
EINECS: 203-534-4
FN: 3261
ADR: 8,III
Merck Indeks: 14,02159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00ffd9383f1f7bdf99ac70a1e45dce0006e0898e&vid=00ffd9383f1f7bdf99ac70a1e45dce0006e0898e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 38.0 to 42.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR