3-Chlorothioanisole ≥96.0% (by GC)

Leverandør: TCI
Warning

Synonymer: (3-Chlorophenyl)(methyl)sulphane, 3-Chlorophenyl methyl sulphide, 1-Chloro-3-(methylthio)benzene

C2070-25G C2070-5G
TCIAC2070-25GEA 970 NOK
TCIAC2070-25G TCIAC2070-5G
3-Chlorothioanisole ≥96.0% (by GC)
3-Chlorothioanisole
Formel: C₇H₇ClS
MW: 158,65 g/mol
Kokepunkt: 244°C
Tetthet: 1,206 g/cm³ (25 °C)
Flash punkt: 106°C
MDL nummer: MFCD00040127
CAS-nummer: 4867-37-2
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01007a503cd3132bd2db904097f596eec529aad3&vid=01007a503cd3132bd2db904097f596eec529aad3&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR