3'-Methylacetophenone ≥97.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Methyl m-tolyl ketone, 1-(m-Tolyl)ethanone, m-Methylacetophenone, 3′-Methylacetophenone

M0191-25ML M0191-100ML
TCIAM0191-25MLEA 675 NOK
TCIAM0191-25ML TCIAM0191-100ML
3'-Methylacetophenone ≥97.0% (by GC)
3'-Methylacetophenone
Formel: C₉H₁₀O
MW: 134,18 g/mol
Kokepunkt: 219°C
Flash punkt: 85°C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00008742
CAS-nummer: 585-74-0
EINECS: 209-561-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a28a876814720aa09991a0c09e575b60f9119025&vid=a28a876814720aa09991a0c09e575b60f9119025&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR