Hexadecan-16-olide ≥97.0% (by GC)
Leverandør: TCI
|
Warning
|
Synonymer:
Oxa-2-cycloheptadecanone, Hexadecanolactone, Oxacycloheptadecan-2-one, 16-Hexadecanolide
H1219-25G
H1219-5G
TCIAH1219-25GEA
2650
NOK
TCIAH1219-25G
TCIAH1219-5G
Hexadecan-16-olide ≥97.0% (by GC)
Hexadecan-16-olide
Formel:
C₁₆H₃₀O₂ MW: 254,41 g/mol Kokepunkt: 188°C Lagringstemperatur: Fryseskap |
MDL nummer:
MFCD00039668 CAS-nummer: 109-29-5 EINECS: 203-662-0 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...