3-Benzoylpyridine ≥99.0% (by GC)

Leverandør: TCI
Warning

Synonymer: Phenyl 3-pyridyl ketone, Phenyl(pyridin-3-yl)methanone

B0305-25G B0305-5G
TCIAB0305-25GEA 1850 NOK
TCIAB0305-25G TCIAB0305-5G
3-Benzoylpyridine ≥99.0% (by GC)
3-Benzoylpyridine
Formel: C₁₂H₉NO
MW: 183,21 g/mol
Kokepunkt: 307°C
Smeltepunkt: 42 °C
MDL nummer: MFCD00006394
CAS-nummer: 5424-19-1
EINECS: 226-561-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58c2a1b1a2bba7b70c2b52a7c5bea443929d787a&vid=58c2a1b1a2bba7b70c2b52a7c5bea443929d787a&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 99.0 %
Melting point 37.0 to 40.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR