3'-Nitroacetophenone ≥98.0% (by GC)
Leverandør: TCI
Synonymer:
m-Nitroacetophenone
N0109-25G
N0109-100G
TCIAN0109-25GEA
220
NOK
TCIAN0109-25G
TCIAN0109-100G
3'-Nitroacetophenone ≥98.0% (by GC)
3'-Nitroacetophenone
Formel:
C₈H₇NO₃ MW: 165,15 g/mol Kokepunkt: 202°C Smeltepunkt: 77…78 °C Tetthet: 1,433 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00007259 CAS-nummer: 121-89-1 EINECS: 204-504-3 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Melting point | 77.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...