3-Aminobenzamide ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Warning

A0630-25G A0630-5G
TCIAA0630-25GEA 1500 NOK
TCIAA0630-25G TCIAA0630-5G
3-Aminobenzamide ≥98.0% (by titrimetric analysis)
3-Aminobenzamide
Formel: C₇H₈N₂O
Kokepunkt: 341 °C (1013 hPa)
Smeltepunkt: 111…115 °C
MDL nummer: MFCD00007989
CAS-nummer: 3544-24-9
EINECS: 222-586-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33d63c5f7d57a58b0eb36cfecb2c1fb1d0b0909b&vid=33d63c5f7d57a58b0eb36cfecb2c1fb1d0b0909b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR