3-Methylcinnamic acid ≥98.0% (by titrimetric analysis)
Leverandør: TCI
Warning
Synonymer: 3-(m-Tolyl)acrylic acid
M1298-25G M1298-5G
TCIAM1298-25GEA 3950 NOK
TCIAM1298-25G TCIAM1298-5G
3-Methylcinnamic acid ≥98.0% (by titrimetric analysis)
3-Methylcinnamic acid
Formel: C₁₀H₁₀O₂
MW: 162,19 g/mol
Kokepunkt: 303 °C (1013 hPa)
Smeltepunkt: 119…120 °C
MDL nummer: MFCD00016844
CAS-nummer: 3029-79-6
EINECS: 221-199-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=56ff02c19ce1371a2a49ed5e7fc9af49b8a94753&vid=56ff02c19ce1371a2a49ed5e7fc9af49b8a94753&prid=web&clid=SAPDAM
Bestill nå
Spesifikasjon Testresultater
Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 116.0 to 120.0 deg-C