3-Nitrocinnamic acid ≥98.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
Synonymer:
3-(3-Nitrophenyl)acrylic acid
N0354-25G
N0354-5G
TCIAN0354-25GEA
745
NOK
TCIAN0354-25G
TCIAN0354-5G
3-Nitrocinnamic acid ≥98.0% (by GC, titration analysis)
3-Nitrocinnamic acid
Formel:
C₉H₇NO₄ MW: 193,160 g/mol |
MDL nummer:
MFCD00007277 CAS-nummer: 555-68-0 EINECS: 209-104-2 Merck Indeks: 14,06595 |
Spesifikasjon Testresultater
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 203.0 to 206.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...