Isophthalic acid ≥99.0% (by GC, titration analysis)
Leverandør: TCI
|
Warning
|
I0155-25G
I0155-500G
TCIAI0155-25GEA
230
NOK
TCIAI0155-25G
TCIAI0155-500G
Isophthalic acid ≥99.0% (by GC, titration analysis)
Isophthalic acid
Formel:
C₈H₆O₄ MW: 166,13 g/mol Kokepunkt: 119 - 121 °C (6 -rr) Smeltepunkt: 340°C Tetthet: 1,54 g/cm³ (20 °C) Lagringstemperatur: Romtemperatur |
MDL nummer:
MFCD00002516 CAS-nummer: 121-91-5 EINECS: 204-506-4 Merck Indeks: 14,05197 |
Spesifikasjon Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Solubility in 1mol/L NaOH | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...