6-Fluoro-1-indanone ≥98.0% (by GC)
Leverandør: TCI
|
Warning
|
F0668-5G
F0668-1G
TCIAF0668-5GEA
2400
NOK
TCIAF0668-5G
TCIAF0668-1G
6-Fluoro-1-indanone ≥98.0% (by GC)
6-Fluoro-1-indanone
Formel:
C₉H₇FO MW: 150,15 g/mol |
MDL nummer:
MFCD01318147 CAS-nummer: 1481-32-9 |
Spesifikasjon Testresultater
Appearance | Light yellow to Brown to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...